MilliporeSigma
  • Chemiluminescence microfluidic system of gold nanoparticles enhanced luminol-silver nitrate for the determination of vitamin B12.
Chemiluminescence microfluidic system of gold nanoparticles enhanced luminol-silver nitrate for the determination of vitamin B12.
Biomedical microdevices (2012-10-12)
Mohammad Kamruzzaman, Al-Mahmnur Alam, Kyung Min Kim, Sang Hak Lee, Young Ho Kim, A N M Hamidul Kabir, Gyu-Man Kim, Trung Dung Dang
ABSTRACT
A rapid and sensitive chemiluminescence (CL) system coupled with a microfluidic chip has been presented to determine vitamin B12 (VB12) based on the reaction of luminol and silver nitrate (AgNO(3)) in the presence of gold nanoparticles (AuNPs). A microfluidic chip was fabricated by a soft-lithographic procedure using polydimethyl siloxane (PDMS) having four inlets and one outlet with a 200 μm wide, 250 μm deep, and 100 mm long microchannel. Ag(+) was used as a chemiluminogenic oxidant in this CL reaction which oxidized luminol to produce strong CL signal in the presence of AuNPs. Luminol reacted with AgNO(3) under the catalysis of AuNPs to produce luminol radicals which reacted with dissolved oxygen and emitted CL light. The proposed CL system was applied to determine the amount of VB12 in VB12 tablets and multivitamin. Under the optimum conditions, the CL intensity of the system was increased with the concentration of VB12 in the range of 0.25-100 ng mL(-1) with the correlation coefficient of 0.9982. The limit of detection was found to be 0.04 ng mL(-1) with the relative standard deviation of 1.56 % for five replicate determinations of 25 ng mL(-1) of VB12. The CL reaction mechanism was demonstrated by UV-visible spectra and CL emission spectra.
Sigma-Aldrich
Silver standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Ag, 1000 ppm Ag
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Silver nitrate solution, 0.05 M
Sigma-Aldrich
Silver nitrate solution, 0.0282 M