MilliporeSigma
  • Loss of Slc4a1b chloride/bicarbonate exchanger function protects mechanosensory hair cells from aminoglycoside damage in the zebrafish mutant persephone.
Loss of Slc4a1b chloride/bicarbonate exchanger function protects mechanosensory hair cells from aminoglycoside damage in the zebrafish mutant persephone.
PLoS genetics (2012-10-17)
Dale W Hailey, Brock Roberts, Kelly N Owens, Andrew K Stewart, Tor Linbo, Remy Pujol, Seth L Alper, Edwin W Rubel, David W Raible
ABSTRACT
Mechanosensory hair cell death is a leading cause of hearing and balance disorders in the human population. Hair cells are remarkably sensitive to environmental insults such as excessive noise and exposure to some otherwise therapeutic drugs. However, individual responses to damaging agents can vary, in part due to genetic differences. We previously carried out a forward genetic screen using the zebrafish lateral line system to identify mutations that alter the response of larval hair cells to the antibiotic neomycin, one of a class of aminoglycoside compounds that cause hair cell death in humans. The persephone mutation confers resistance to aminoglycosides. 5 dpf homozygous persephone mutants are indistinguishable from wild-type siblings, but differ in their retention of lateral line hair cells upon exposure to neomycin. The mutation in persephone maps to the chloride/bicarbonate exchanger slc4a1b and introduces a single Ser-to-Phe substitution in zSlc4a1b. This mutation prevents delivery of the exchanger to the cell surface and abolishes the ability of the protein to import chloride across the plasma membrane. Loss of function of zSlc4a1b reduces hair cell death caused by exposure to the aminoglycosides neomycin, kanamycin, and gentamicin, and the chemotherapeutic drug cisplatin. Pharmacological block of anion transport with the disulfonic stilbene derivatives DIDS and SITS, or exposure to exogenous bicarbonate, also protects hair cells against damage. Both persephone mutant and DIDS-treated wild-type larvae show reduced uptake of labeled aminoglycosides. persephone mutants also show reduced FM1-43 uptake, indicating a potential impact on mechanotransduction-coupled activity in the mutant. We propose that tight regulation of the ionic environment of sensory hair cells, mediated by zSlc4a1b activity, is critical for their sensitivity to aminoglycoside antibiotics.
Product Description
Supelco
Neomycin trisulfate salt hydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, suitable for plant cell culture
Neomycin sulphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kanamycin disulfate salt from Streptomyces kanamyceticus, aminoglycoside antibiotic
Sigma-Aldrich
Neomycin solution, with 10 mg/mL neomycin in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture