MilliporeSigma
  • [The study of corrective role of plant--based drugs with antioxidant action in experimental intoxication with potassium dichromate].
[The study of corrective role of plant--based drugs with antioxidant action in experimental intoxication with potassium dichromate].
Gigiena i sanitariia (2012-10-24)
A N Zinalieva, T K Karimov, S K Bermagambetova
ABSTRACT
The disturbance of the structure and function of pulmonary surfactant system (pSS) is an essential component of almost all pulmonary diseases. Chromium compounds have a damaging effect on the PSS, which contributes to the formation of nonspecific pulmonary diseases and a burden of chrome disease. The results showed that the oil of licorice (Glycyrrhiza glabra L) protects PSS in chronic chrome intoxication.
Sigma-Aldrich
Chromium standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Cr, 1000 ppm Cr
Sigma-Aldrich
Potassium dichromate solution, 1/60 M
Sigma-Aldrich
Potassium dichromate, JIS special grade, ≥99.5%
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Supelco
Potassium dichromate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%