MilliporeSigma
  • Preparation, characterization and efficacy evaluation of synthetic biocompatible polymers linking natural antioxidants.
Preparation, characterization and efficacy evaluation of synthetic biocompatible polymers linking natural antioxidants.
Molecules (Basel, Switzerland) (2012-10-30)
Sonia Trombino, Roberta Cassano, Teresa Ferrarelli, Sonia Leta, Francesco Puoci, Nevio Picci
ABSTRACT
The purpose of this work was the synthesis, characterization and efficacy evaluation of new biocompatible antioxidant polymers linking trans-ferulic acid or a-lipoic acid. In particular, ferulic or lipoic acid were introduced in the preformed polymeric backbone. The new antioxidant biopolymers were characterized by Fourier transform infrared spectroscopy and gel permeation chromatography. The degree of functionalization (moles of antioxidant per gram of polymer) was determined by the Gaur-Gupta method for free amino group determination and by the Folin method for the phenolic groups. Their ability to inhibit lipid peroxidation were estimated in rat liver microsomal membranes induced in vitro by tert-BOOH (tert-butyl hydroperoxide), as a source of free radicals. The DPPH (1,1-diphenyl-2-picrylhydrazyl) radical-scavenging effect was also evaluated. The obtained systems, with different solubility, showed strong antioxidant and antiradical activities, suggesting potential use as packaging materials for foods, cosmetics, pharmaceuticals and personal care products. Moreover, the cytotoxicity of the synthesized polymers was also evaluated on Caco-2 cell cultures in order to verify their biocompatibility when exposed to an absorptive epithelial cell line.
Sigma-Aldrich
N,N-Dimethylacrylamide, 99%, contains 500 ppm monomethyl ether hydroquinone as inhibitor
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-Ferulic acid, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Polyacrylamide, average Mn 150,000