MilliporeSigma
  • Contrasting binding of fisetin and daidzein in γ-cyclodextrin nanocavity.
Contrasting binding of fisetin and daidzein in γ-cyclodextrin nanocavity.
Journal of photochemistry and photobiology. B, Biology (2012-11-28)
Biswapathik Pahari, Bidisha Sengupta, Sandipan Chakraborty, Briannica Thomas, Dyffreyon McGowan, Pradeep K Sengupta
ABSTRACT
Steady state and time resolved fluorescence along with anisotropy and induced circular dichroism (ICD) spectroscopy provide useful tools to observe and understand the behavior of the therapeutically important plant flavonoids fisetin and daidzein in γ-cyclodextrin (γ-CDx) nanocavity. Benesi-Hildebrand plots indicated 1:1 stoichiometry for both the supramolecular complexes. However, the mode of the binding of fisetin significantly differs from daidzein in γ-CDx, as is observed from ICD spectra which is further confirmed by docking studies. The interaction with γ-CDx proceeds mainly by the phenyl ring and partly by the chromone ring of fisetin whereas only the phenyl ring takes part for daidzein. A linear increase in the aqueous solubility of the flavonoids is assessed from the increase in the binding of the flavonoids with the γ-CDx cavity, which are determined by the gradual increase in the ICD signal, fluorescence emission as well as increase in fluorescence anisotropy with increasing (γ-CDx). This confirms γ-CDx as a nanovehicle for the flavonoids fisetin and daidzein in improving their bioavailability.
Sigma-Aldrich
γ-Cyclodextrin, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥90.0% cyclodextrin basis (HPLC)
Supelco
Daidzein, analytical standard
Sigma-Aldrich
Daidzein, ≥98%, synthetic