MilliporeSigma
  • Effect of ionic liquid pretreatment on the composition, structure and biogas production of water hyacinth (Eichhornia crassipes).
Effect of ionic liquid pretreatment on the composition, structure and biogas production of water hyacinth (Eichhornia crassipes).
Bioresource technology (2012-11-29)
Jing Gao, Li Chen, Zongcheng Yan, Lin Wang
ABSTRACT
The effect of the pretreatment of water hyacinth with ionic liquid and co-solvent on the lignocellulosic composition, structural change and biogas production was evaluated in this study. The results from regenerated water hyacinth indicate that, the content of the lignocellulosic composition was changed, the crystallinity of the structure was decreased, and the surface became more porous. After the pretreatment with 1-N-butyl-3-methyimidazolium chloride ([Bmim]Cl)/dimethyl sulfoxide (DMSO) under 120°C for 120min, the cellulose content of regenerated water hyacinth was increased by 27.9%, 49.2% of the lignin was removed, and the biogas yield was increased by 97.6% as compared with unpretreated water hyacinth. The ionic liquids and co-solvents were successfully recovered by forming aqueous biphasic systems with K3PO4.
Sigma-Aldrich
Phosphorus standard solution, 1000 ppm P
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution