MilliporeSigma
  • Toxicity, feeding rate and growth rate response to sub-lethal concentrations of anthracene and benzo [a] pyrene in milkfish Chanos chanos (Forskkal).
Toxicity, feeding rate and growth rate response to sub-lethal concentrations of anthracene and benzo [a] pyrene in milkfish Chanos chanos (Forskkal).
Bulletin of environmental contamination and toxicology (2012-12-01)
L Palanikumar, A K Kumaraguru, C M Ramakritinan, M Anand
ABSTRACT
The feeding rate, growth rate and gross conversion efficiency were studied in milkfish Chanos chanos for 28 days of exposure to sub-lethal concentrations of anthracene (1.00, 2.00, 3.00, 6.00 and 12.0 μg l(-1)) and benzo [a] pyrene (0.30, 0.70, 1.40, 2.80 and 5.60 μg l(-1)) under continuous flow through bioassays. Based on survival and growth data, No Observed Effect Concentration; Lowest Observed Effect Concentration were estimated after 28 days, the values for anthracene were 2.03 and 3.09 μg l(-1), and the values for benzo [a] pyrene were 0.82 and 1.46 μg l(-1), respectively. Anthracene and benzo [a] pyrene exposure caused reduction in feeding and growth rate.
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland