MilliporeSigma
Role of melatonin and luzindole in rat mammary cancer.
Journal of investigative surgery : the official journal of the Academy of Surgical Research (2012-12-12)
Ugurlu M Umit, Terzioglu Berna, Kaya Handan, Erbarut Ipek, Yegen Berrak, Erzik Can, Gulluoglu M Bahadir
ABSTRACT
Recent studies have analyzed the efficacy of various agents in experimental chemoprevention trials. In our study, the effects of melatonin (Mel) and its antagonist Luzindole (Luz) on Heme oxygenase-1 (HO-1) in a NMU (N-methyl-N-nitrosourea)-induced rat mammary carcinoma model are investigated. We aim to demonstrate the relationship between Mel and HO-1. Spraque-Dawley rats were treated with NMU at age 55 days to induce mammary carcinoma. Forty-eight rats were divided into four groups consisting of: (a) physiological saline group (PSG); (b) control group, NMU is given; (c) Mel group (500 μg daily); (d) Mel antagonist Luz group (0.25 mg/kg/day i.p.). The animals were sacrificed; their serum and tissues were sampled for histopathologic evaluation, markers of endocrine derangement (serum prolactin, estradiol, and progesterone levels), apoptotic changes, DNA fragmentation, markers of oxidative stress and HO-1 immune expression were measured. Most tumors developed in the Luz group (42%), followed by the control group (33%), and the Mel group (17%). The tumor latency was longer in Mel-treated group (control and Luz at week 17, Mel at week 21). The maximum tumor volume was also smaller in Mel group when compared to control and Luz groups (p < .05). In Mel group estradiol, progesterone, and prolactin levels were decreased compared to control group (p < .001; p < .01; and p < .01) and levels of apoptotic activity and DNA fragmentation ratio increased. The increment of HO-1 expression with Mel is described; possible underlying mechanisms of these effects await further investigations.
Sigma-Aldrich
Luzindole, ≥90%