MilliporeSigma
  • Yttrium phosphasalen initiators for rac-lactide polymerization: excellent rates and high iso-selectivities.
Yttrium phosphasalen initiators for rac-lactide polymerization: excellent rates and high iso-selectivities.
Journal of the American Chemical Society (2012-12-13)
Clare Bakewell, Thi-Phuong-Anh Cao, Nicholas Long, Xavier F Le Goff, Audrey Auffrant, Charlotte K Williams
ABSTRACT
Highly active yttrium phosphasalen initiators for the stereocontrolled ring-opening polymerization of rac-lactide are reported. The initiators are coordinated by a new class of ancillary ligand: an iminophosphorane derivative of the popular "salen" ligand, termed "phosphasalen". Changing the phosphasalen structure enables access to high iso-selectivities (P(i) = 0.84) or hetero-selectivities (P(s) = 0.87). The initiators also show very high rates, excellent polymerization control, and tolerance to low loadings; furthermore, no chiral auxiliaries/ligands are needed for the stereocontrol. The combination of such high rates with high iso-selectivities is very unusual.
Sigma-Aldrich
N,N′-Bis(salicylidene)ethylenediamine, 98%
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%