MilliporeSigma
  • Chromogenic substrate from 4-nitro-1-naphthol for hydrolytic enzyme of neutral or slightly acidic optimum pH: 4-nitro-1-naphthyl-β-D-galactopyranoside as an example.
Chromogenic substrate from 4-nitro-1-naphthol for hydrolytic enzyme of neutral or slightly acidic optimum pH: 4-nitro-1-naphthyl-β-D-galactopyranoside as an example.
Bioorganic & medicinal chemistry letters (2012-12-27)
Jizheng Dang, Hongbo Liu, Xiaolan Yang, Yi Zhang, Yanling Xie, Yuanli Li, Jun Pu, Juan Liao, Yonghua Yuan, Fei Liao
ABSTRACT
At pH from 5.5 to 7.6, absorptivity of 4-nitro-1-naphthol at 450 nm is over 2.1-fold of that of para-nitrophenol at 405 nm and over 9.6-fold of that of ortho-nitrophenol at 415 nm. On 4-nitro-1-naphthyl-β-D-galactopyranoside at pH 7.4, catalytic efficiency of Escherichia coli β-D-galactosidase is 3-fold of that on para-nitrophenyl-β-D-galactopyranoside and about 40% of that on ortho-nitrophenyl-β-D-galactopyranoside, and produces a lower quantification limit of penicillin G by enzyme-linked-immunoabsorbent-assay. Hence, 4-nitro-1-naphthol is favorable to prepare chromogenic substrates of hydrolytic enzymes of neutral or slightly acidic optimum pH.
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Sigma-Aldrich
1-Naphthol, SAJ special grade, ≥99.0%
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Penicillin G potassium salt
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%