MilliporeSigma
  • Effects of mono-(2-ethylhexyl) phthalate (MEHP) on chicken germ cells cultured in vitro.
Effects of mono-(2-ethylhexyl) phthalate (MEHP) on chicken germ cells cultured in vitro.
Environmental science and pollution research international (2013-01-29)
Edith Guibert, Bérénice Prieur, Ronan Cariou, Frédérique Courant, Jean Philippe Antignac, Bertrand Pain, Jean Pierre Brillard, Pascal Froment
ABSTRACT
In recent decades, many toxicological tests based on in vivo or in vitro models, mainly from mammalian (rat-mouse) and fish species, were used to assess the risks raised by contact or ingestion of molecules of pharmaceutical, agricultural, or natural origin. But no, or few, in vitro tests using other non-mammalian models such as bird have been explored despite their advantages: the embryonic gonads of birds have a high plasticity of development sensitive to estrogen, and sperm production is nearly two times faster than in rodents. Hence, we have established an in vitro culture of germ cells and somatic cells from chicken post-natal testis, and we have evaluated the sensitivity against the endocrine disruptor compound mono-(2-ethylhexyl) phthalate (MEHP) in comparison to previous studies using rodent and human models. After 96 h of exposure in presence of 10 μM MEHP, chicken seminiferous tubules cultures present a structural alteration, a reduction in cell proliferation and in germ cells population. Apoptosis of germ and somatic cells increases in presence of 1 μM MEHP. Furthermore, MEHP does not affect inhibin B and lactate production by Sertoli cells. These results are in accordance with previous studies using rat, mice, or human culture of testicular cells and in similar range of exposures or even better sensitivity for some "end-points" (biological parameters). In conclusion, the establishment of this postnatal testicular cells culture could be considered as an alternative method to in vivo experiments frequently used for evaluating the impact on the terrestrial wildlife species. This method could be also complementary to mammal model due to the limiting number of animals used and its elevated sensitivity.
Product Description
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00