MilliporeSigma
Effects of topicals on the aging skin process.
Facial plastic surgery clinics of North America (2013-02-02)
J Regan Thomas, Tatiana K Dixon, Tapan K Bhattacharyya
ABSTRACT
This article summarizes the antiaging properties of retinoids, glycolic acid, ascorbic acid, and peptide topicals. The supporting evidence is taken from the literature and the primary author's research, consisting of previously published data and new results from ongoing projects.
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%