MilliporeSigma
Biological functions of iduronic acid in chondroitin/dermatan sulfate.
The FEBS journal (2013-02-28)
Martin A Thelin, Barbara Bartolini, Jakob Axelsson, Renata Gustafsson, Emil Tykesson, Edgar Pera, Åke Oldberg, Marco Maccarana, Anders Malmstrom
ABSTRACT
The presence of iduronic acid in chondroitin/dermatan sulfate changes the properties of the polysaccharides because it generates a more flexible chain with increased binding potentials. Iduronic acid in chondroitin/dermatan sulfate influences multiple cellular properties, such as migration, proliferation, differentiation, angiogenesis and the regulation of cytokine/growth factor activities. Under pathological conditions such as wound healing, inflammation and cancer, iduronic acid has diverse regulatory functions. Iduronic acid is formed by two epimerases (i.e. dermatan sulfate epimerase 1 and 2) that have different tissue distribution and properties. The role of iduronic acid in chondroitin/dermatan sulfate is highlighted by the vast changes in connective tissue features in patients with a new type of Ehler-Danlos syndrome: adducted thumb-clubfoot syndrome. Future research aims to understand the roles of the two epimerases and their interplay with the sulfotransferases involved in chondroitin sulfate/dermatan sulfate biosynthesis. Furthermore, a better definition of chondroitin/dermatan sulfate functions using different knockout models is needed. In this review, we focus on the two enzymes responsible for iduronic acid formation, as well as the role of iduronic acid in health and disease.
Product Description
Chondroitin sulfate sodium, European Pharmacopoeia (EP) Reference Standard
Chondroitin sulfate sodium (marine), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Chondroitin 4-sulfate sodium salt from bovine trachea, mixture of isomers, main component: chondroitin 4-sulfate