MilliporeSigma
  • Effects of pyrethroid resistance on the cost effectiveness of a mass distribution of long-lasting insecticidal nets: a modelling study.
Effects of pyrethroid resistance on the cost effectiveness of a mass distribution of long-lasting insecticidal nets: a modelling study.
Malaria journal (2013-02-28)
Olivier J T Briët, Melissa A Penny, Diggory Hardy, Taiwo S Awolola, Wim Van Bortel, Vincent Corbel, Roch K Dabiré, Josiane Etang, Benjamin G Koudou, Patrick K Tungu, Nakul Chitnis
ABSTRACT
The effectiveness of insecticide-treated nets in preventing malaria is threatened by developing resistance against pyrethroids. Little is known about how strongly this affects the effectiveness of vector control programmes. Data from experimental hut studies on the effects of long-lasting, insecticidal nets (LLINs) on nine anopheline mosquito populations, with varying levels of mortality in World Health Organization susceptibility tests, were used to parameterize malaria models. Both simple static models predicting population-level insecticidal effectiveness and protection against blood feeding, and complex dynamic epidemiological models, where LLINs decayed over time, were used. The epidemiological models, implemented in OpenMalaria, were employed to study the impact of a single mass distribution of LLINs on malaria, both in terms of episodes prevented during the effective lifetime of the batch of LLINs, and in terms of net health benefits (NHB) expressed in disability-adjusted life years (DALYs) averted during that period, depending on net type (standard pyrethroid-only LLIN or pyrethroid-piperonyl butoxide combination LLIN), resistance status, coverage and pre-intervention transmission level. There were strong positive correlations between insecticide susceptibility status and predicted population level insecticidal effectiveness of and protection against blood feeding by LLIN intervention programmes. With the most resistant mosquito population, the LLIN mass distribution averted up to about 40% fewer episodes and DALYs during the effective lifetime of the batch than with fully susceptible populations. However, cost effectiveness of LLINs was more sensitive to the pre-intervention transmission level and coverage than to susceptibility status. For four out of the six Anopheles gambiae sensu lato populations where direct comparisons between standard LLINs and combination LLINs were possible, combination nets were more cost effective, despite being more expensive. With one resistant population, both net types were equally effective, and with one of the two susceptible populations, standard LLINs were more cost effective. Despite being less effective when compared to areas with susceptible mosquito populations, standard and combination LLINs are likely to (still) be cost effective against malaria even in areas with strong pyrethroid resistance. Combination nets are likely to be more cost effective than standard nets in areas with resistant mosquito populations.
Sigma-Aldrich
Piperonylbutoxide, technical grade, 90%
Supelco
Piperonylbutoxide, PESTANAL®, analytical standard