MilliporeSigma
  • First and unexpected synthesis of macrocyclic cyclophane-based unusual alpha-amino acid derivatives by phosphazene base without high dilution conditions.
First and unexpected synthesis of macrocyclic cyclophane-based unusual alpha-amino acid derivatives by phosphazene base without high dilution conditions.
Bioorganic & medicinal chemistry letters (2002-03-23)
Sambasivarao Kotha, Somnath Halder, Lakshminarasimhan Damodharan, Vasantha Pattabhi
ABSTRACT
First synthesis of a macrocylic cyclophane-based unusual alpha-amino acid derivative 11 by coupling of ethyl isocyanoacetate with 1,2-bis(4-bromomethylphenyl)ethane under phase-transfer catalysis (PTC) conditions. Phosphazene base such as 2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine (BEMP) is useful to improve the yield of cyclophane derivative without high dilution conditions.
Sigma-Aldrich
2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, purum, ≥98.0% (GC)