MilliporeSigma
  • Adaptive microbial population shifts in response to a continuous ethanol blend release increases biodegradation potential.
Adaptive microbial population shifts in response to a continuous ethanol blend release increases biodegradation potential.
Environmental pollution (Barking, Essex : 1987) (2013-05-01)
Jie Ma, Carlos W Nossa, Zongming Xiu, William G Rixey, Pedro J J Alvarez
ABSTRACT
The fate of fuel releases largely depends on the poorly-understood response in microbial community structure and function. Here, we evaluate the impacts to the microbial community resulting from a pilot-scale continuous release (10 months) of a 10% v:v ethanol solution mixed with benzene and toluene (50 mg/L each). Microbial population shifts were characterized by pyrosequencing-based 16S rRNA analysis and by quantitative PCR targeting Bacteria, Archaea, and functional genes for methanogenesis (mcrA), acetogenesis (fhs) and aerobic degradation of aromatic hydrocarbons (PHE), which could occur in hypoxic micro-environments. The release stimulated microbial growth, increased species richness and diversity, and selected for genotypes involved in fermentative degradation (the relative abundance of mcrA and fhs increased 18- and 6-fold, respectively). The growth of putative hydrocarbon degraders and commensal anaerobes, and increases in microbial diversity and in degradation rates suggest an adaptive response that increases the potential for natural attenuation of ethanol blend releases.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Toluene, HPLC grade, 99.8%
Supelco
Benzene, analytical standard
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%