MilliporeSigma
  • The β-fibrinogen -455G/A gene polymorphism and the risk of ischaemic stroke in a Polish population.
The β-fibrinogen -455G/A gene polymorphism and the risk of ischaemic stroke in a Polish population.
Neurologia i neurochirurgia polska (2013-05-08)
Aleksandra Golenia, Joanna Chrzanowska-Wasko, Jeremiasz Jagiella, Marcin Wnuk, Antoni Ferens, Aleksandra Klimkowicz-Mrowiec, Mateusz Adamski, Irena Ciecko-Michalska, Agnieszka Slowik
ABSTRACT
Ischaemic stroke is considered to be multifactorial and interactions between environmental and genetic factors play an important role. Although vascular risk factors are well known, the genetic ones are still undiscovered. In the present study, we assessed the significance of the β-fibrinogen -455G/A gene polymorphism and the risk of ischaemic stroke in a Polish population. 426 ischaemic stroke patients classified according to stroke aetiologies (small vessel disease, large vessel disease or cardioembolic stroke) and 234 controls were included in the study. The association of the β-fibrinogen genotypes with ischaemic stroke was tested using logistic regression analysis under dominant, recessive or additive models of inheritance. The allele and genotype distributions of the β-fibrinogen -455G/A gene polymorphism did not differ significantly between patients and controls (patients: G - 75%, GG - 56.6%, GA - 36.8%, AA - 6.6%; controls: G - 73.7%, GG - 57.3%, GA - 32.9%, AA - 9.8%; p > 0.05, χ2). In addition, logistic regression analysis adjusted for the known risk factors, i.e. hypertension, ischaemic heart disease, myocardial infarction, hypercholesterolaemia, diabetes mellitus and smoking, did not show a role of the studied polymorphism in ischaemic stroke. The β-fibrinogen -455G/A gene polymorphism is not a risk factor for ischaemic stroke in a Polish population.
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00