Merck
[Dipentafluorophenyl carbonate--a reagent for peptide synthesis].
Bioorganicheskaia khimiia (1989-04-01)
V N Medvedkin, Mitin YuV, L V Klimenko, N N Podgornova, A I Bystrichenko
ABSTRACT
Dipentafluorophenylcarbonate, belonging to transesterifiying reagents, has been prepared and used for the synthesis of pentafluorophenyl esters of amino acids. In contrast to many other reagents of the kind, its preparation is simple, it is highly reactive and at the same time stable upon storage.
Sigma-Aldrich
Bis(pentafluorophenyl) carbonate, 97%