MilliporeSigma
  • [Studies on the pharmaceutical availability of chlortetracycline hydrochloride in selected ointment bases].
[Studies on the pharmaceutical availability of chlortetracycline hydrochloride in selected ointment bases].
Acta poloniae pharmaceutica (1989-01-01)
M Gadomska-Nowak, H Wojdak, J Godlewska, J Pasich
ABSTRACT
Influence of non-ionic tensides on liberation of chlorotetracycline hydrochloride from ointment bases was investigated. Non-ionic tensides were found to enhance liberation of chlorotetracycline hydrochloride from lipophilic ointment bases, while liberation of this antibiotic from hydrophilic ointment bases proved to be depressed.
Sigma-Aldrich
Sorbitan monopalmitate
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00