MilliporeSigma
Some physical and chemical properties of Bhanja virus.
Acta virologica (1986-09-01)
Z Hubálek
ABSTRACT
Bhanja virus is acid-labile, relatively thermostable, resistant to trypsin and heparin; a complete inactivation was achieved with chloramine B or formaldehyde, while phenol was ineffective, and UV radiation only partially effective.
Sigma-Aldrich
N-Chlorobenzenesulfonamide sodium salt, ~28% active chlorine basis