MilliporeSigma
  • Synthesis and pharmacological evaluation of N,N-di-n-propyldopamine congeners containing phenolic bioisosteres.
Synthesis and pharmacological evaluation of N,N-di-n-propyldopamine congeners containing phenolic bioisosteres.
Journal of pharmaceutical sciences (1987-05-01)
R D Clark, J M Caroon, N E Isaac, D L McClelland, A D Michel, T A Petty, R P Rosenkranz, L D Waterbury
ABSTRACT
A series of analogues of N,N-di-n-propyldopamine (DPDA) in which the 3-hydroxyl group was replaced by bioisosteric groups was prepared and evaluated for D1- and D2-receptor affinity. The 3-methane-sulfonamide analogue (18) had a higher affinity for the D2 receptor than DPDA and was more selective for the D2 receptor. The 3-formamide derivative (15) also retained significant D2 affinity. Both of these compounds demonstrated in vivo cardiovascular and renal profiles in an anesthetized rat model that were consistent with selective D2-receptor agonism.
Sigma-Aldrich
Methanesulfonamide, ≥97.0% (CHN)