MilliporeSigma
An improved simple procedure for the determination of thiamine in food.
Die Nahrung (1986-01-01)
B Sebecić, I Vedrina-Dragojević
ABSTRACT
A modified fluorometric method for the determination of thiamine in food is described. The procedure is simple, reproducible, does not demand costly outfit and chemicals and is not susceptible to interference. The recovery of added thiamine is good.
Product Description
Supelco
Thiochrome, analytical standard