MilliporeSigma
  • A convenient protein substrate for the determination of protease specificity: reduced and S-3-(trimethylated amino)propylated lysozyme.
A convenient protein substrate for the determination of protease specificity: reduced and S-3-(trimethylated amino)propylated lysozyme.
Analytical biochemistry (1985-02-15)
K Okazaki, T Imoto, H Yamada
ABSTRACT
Reduced lysozyme was alkylated with 3-bromopropyltrimethylammonium bromide to give reduced and S-3-(trimethylated amino)propylated lysozyme. It was soluble in a wide range of pH and is suitable as the protein substrate to determine protease specificity.
Sigma-Aldrich
(3-Bromopropyl)trimethylammonium bromide, 97%