MilliporeSigma
  • Inactivation of horse liver alcohol dehydrogenase by modification of cysteine residue 174 with 3-bromopropionic acid.
Inactivation of horse liver alcohol dehydrogenase by modification of cysteine residue 174 with 3-bromopropionic acid.
Biochemistry (1984-01-17)
V K Chadha, B V Plapp
ABSTRACT
Horse liver alcohol dehydrogenase is inactivated with Michaelis kinetics at pH 7 and 25 degrees C by 3-bromopropionic acid. In the absence of NAD+, the Ki is 2 mM, and the pseudo bimolecular rate constant (k3/Ki) is 0.03 M-1 s-1; in the presence of 1 mM NAD+, Ki is 2.3 mM, and k3/Ki is 0.006 M-1 s-1. 3-Bromopropionic acid is a competitive inhibitor, Ki of 0.4 mM, against ethanol as a substrate. Inactivation was prevented in the ternary complexes with NAD+ X pyrazole and NADH X isobutyramide, was retarded by NAD+, NADH, or bipyridine, and was almost unaffected by imidazole and AMP. Carboxyethylated enzyme did not detectably (as observed spectrophotometrically) bind bipyridine, NAD+, or NADH. Enzyme was inactivated with radioactive 3-bromopropionic acid, aminoethylated, and digested with trypsin and chymotrypsin. Analysis of the labeled peptides showed that Cys-174 was predominantly modified. In the presence of 1 mM NAD+, the reaction was much less specific. The interaction of the carboxyl group of 3-bromopropionic acid with the guanidino group of Arg-369 probably facilitates the selective reaction with Cys-174, which is ligated to the zinc at the active site. Carboxyethylation apparently inactivates by interfering with the proper binding of the pyrophosphate of the coenzyme to the enzyme.
Sigma-Aldrich
3-Bromopropionic acid, 97%