MilliporeSigma
  • [Effect of isatin and its derivatives on some indicators of carbohydrate metabolism].
[Effect of isatin and its derivatives on some indicators of carbohydrate metabolism].
Voprosy meditsinskoi khimii (1984-09-01)
V N Garalene, L I Mazhilis
ABSTRACT
Effect of isatin and of three its derivatives on content of lactate, pyruvate and glycogen was studied in rat tissues. The substances studied, except of 5-butylisatin, decreased the content of lactic acid with simultaneous increase of glycogen content in liver tissue under normal conditions. The pyruvate content was most markedly altered in presence of 5-bromoisatin and 5-butylisatin. In deficiency of oxygen a distinct increase in content of lactate and pyruvate was noted in animal tissues after the 5-bromoisatin preadministration. Under conditions of oxygen deficiency the substances studied were of distinct significance for more economical utilization of glycogen in liver tissue and skeletal muscles.
Sigma-Aldrich
5-Bromoisatin, technical grade, 90%