MilliporeSigma
  • Thyroid xanthine oxidase and its role in thyroid iodine metabolism in the rat: difference between effects of allopurinol and tungstate.
Thyroid xanthine oxidase and its role in thyroid iodine metabolism in the rat: difference between effects of allopurinol and tungstate.
The Journal of endocrinology (1982-10-01)
J Kawada, Y Shirakawa, Y Yoshimura, M Nishida
ABSTRACT
The role of xanthine oxidase in thyroid function was studied in the rat in vivo by different approaches. Allopurinol, an inhibitor of xanthine oxidase, was administered by mixing it with a powdered diet (16 mg/100 g body wt per day for 10 days). This treatment significantly reduced the total uptake of iodide and inhibited the organification of iodide in the rat thyroid gland. Thyroid xanthine oxidase and dehydrogenase were almost completely inactivated by tungstate, which was given to rats (100 p.p.m./animal per day in drinking water for 10 days) maintained on a purified diet containing low levels of molybdenum. Under these conditions, no inhibitory effect was observed on synthesis of thyroid hormones. It therefore seemed reasonable to assume that the suppressive effect of allopurinol on the biosynthesis of thyroid hormones is not mediated by xanthine oxidase.
Sigma-Aldrich
Tungstic acid, 99%
Sigma-Aldrich
Tungstic acid, ≥98.0% (calcined substance, T), powder