MilliporeSigma
  • Flowering as metamorphosis: two sequential signals regulate floral initiation in Lolium temulentum.
Flowering as metamorphosis: two sequential signals regulate floral initiation in Lolium temulentum.
Development (Cambridge, England) (1996-11-01)
C N McDaniel, L K Hartnett
ABSTRACT
We investigated floral initiation in the long-day monocot Lolium temulentum, strain Ceres, by culturing apices explanted from photoperiodically induced plants at various times after one inductive long day onto medium with, and without, gibberellin. Apices cultured on the first day after the inductive long day usually required gibberellin in the medium to initiate floral morphogenesis while apices explanted on the second day after induction did not require gibberellin. Apices explanted on the first day after induction onto medium without gibberellin grew vegetatively for many days but a several-day exposure to culture medium with gibberellin at any time caused most apices to initiate floral morphogenesis. The gibberellin synthesis inhibitor, ancymidol, when applied to plants before apex excision and when present in the culture medium reduced floral initiation by more than 50% in the absence of added gibberellin in the medium, but it was ineffective in the presence of gibberellin. These results indicated that floral initiation in photoperiodically induced plants resulted from two signals acting at the apex. The first signal induced the apex into a florally determined state and then the second signal, gibberellin, elicited expression of the florally determined state. Leaf removal and culture of apices from plants previously treated with gibberellin provided evidence that the leaf-applied gibberellin did not itself act on the apex to cause floral determination or initiation. Rather, the exogenous gibberellin appeared to stimulate the production of a signal in the leaves that then led to floral initiation.
Sigma-Aldrich
Ancymidol, suitable for plant cell culture, BioReagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15 mg
Estimated to ship on February 05, 2025
Details...
₩212,541