MilliporeSigma
  • Carcinogenicity of methylurea or morpholine in combination with sodium nitrite in rat multi-organ carcinogenesis bioassay.
Carcinogenicity of methylurea or morpholine in combination with sodium nitrite in rat multi-organ carcinogenesis bioassay.
Japanese journal of cancer research : Gann (1997-11-25)
M Kitano, N Takada, T Chen, H Ito, T Nomura, H Tsuda, C P Wild, S Fukushima
ABSTRACT
For carcinogenic risk assessment of combinations of N-nitroso precursors in man, the effects of feeding methylurea (MU) or morpholine (Mor) plus sodium nitrite (NaNO2) were investigated using a multi-organ carcinogenesis model. In experiment 1, to initiate multiple organs, groups of 10 or 20 male F344 rats were treated with 6 carcinogens targeting different organs. Starting a week after completion of this initiation phase, animals were given 0.1% MU or 0.5% Mor in their food and/or 0.15% NaNO2 in their drinking water for 23 weeks. The induction of tumors and/or preneoplastic lesions in the forestomach and esophagus was significantly increased in the group receiving MU plus NaNO2. The numbers and areas of liver glutathione S-transferase placental form (GST-P)-positive foci were significantly elevated with MU or Mor plus NaNO2. Experiment 2 was conducted to assess formation of N-nitroso compounds in the stomach, and to detect DNA adduct generation in target organs by immunohistochemical staining. Groups of 5 or 14 animals were starved overnight, then given 0.4% MU or 2.0% Mor in the diet, or basal diet alone for 1 h. Then NaNO2 or distilled water was given intragastrically. The mean gastric N-methyl-N-nitrosourea yield in the MU plus NaNO2 group was 7700 micrograms at 2 h after combined administration. The mean N-nitrosomorpholine yield in the group given Mor plus NaNO2 was 6720 micrograms. Immunohistochemically, N7-methyldeoxyguanosine-positive nuclei were evident in the forestomach epithelium at 8 h after the combination treatment with MU plus NaNO2.
Sigma-Aldrich
N-Methylurea, 97%