MilliporeSigma
  • Nuclear magnetic resonance spectroscopic analysis of homoallylic and bis homoallylic substituted methyl fatty ester derivatives.
Nuclear magnetic resonance spectroscopic analysis of homoallylic and bis homoallylic substituted methyl fatty ester derivatives.
Lipids (1995-02-01)
M S Jie, K L Cheng
ABSTRACT
Using a combination of selective irradiation 1H nuclear magnetic resonance experiments and two-dimensional 1H-13C correlation spectroscopy spectral analysis of homoallylic and bis homoallylic substituted (azido, acetoxy, chloro and oxo) fatty ester derivatives, the carbon shifts of the ethylenic carbon atoms were determined. In the case of methyl 12-azido-9Z-octadecenoate (homoallylic), the carbon chemical shifts of the ethylenic C-9 and C-10 carbon nuclei are 133.092 and 124.596 ppm, respectively. In methyl 9-azido-12Z-octadecenoate (bis homoallylic), the carbon chemical shift of the ethylenic C-12 and C-13 carbon nuclei are 128.118 and 131.243 ppm, respectively.
Sigma-Aldrich
Methyl ricinoleate, ≥99% (GC)
Supelco
Methyl ricinoleate, analytical standard