MilliporeSigma
  • Powder diffraction study of the hydrogen bonds in nitroxoline and its hydrochloride.
Powder diffraction study of the hydrogen bonds in nitroxoline and its hydrochloride.
Acta crystallographica. Section C, Crystal structure communications (2002-01-10)
A V Yatsenko, K A Paseshnichenko, V V Chernyshev, H Schenk
ABSTRACT
The crystal structures of 8-hydroxy-5-nitroquinoline, C9H6N2O3, (I), and 8-hydroxy-5-nitroquinolinium chloride, C9H7N2O3+*Cl-, (II), have been determined from X-ray powder data. In (I), the molecules are linked via moderately strong hydrogen bonds to form dimers. Such a packing motif is likely to be responsible for the low solubility of (I) in water. In (II), the inversion-related cations form stacks, and anions fill the interstack channels.
Sigma-Aldrich
8-Hydroxy-5-nitroquinoline, 96%