MilliporeSigma
  • [Studies on the identification of psychotropic substances. VIII. Preparation and various analytical data of reference standard of some stimulants, amfepramone, cathinone, N-ethylamphetamine, fenethylline, fenproporex and mefenorex].
[Studies on the identification of psychotropic substances. VIII. Preparation and various analytical data of reference standard of some stimulants, amfepramone, cathinone, N-ethylamphetamine, fenethylline, fenproporex and mefenorex].
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences (1992-01-01)
M Shimamine, K Takahashi, Y Nakahara
ABSTRACT
The Reference Standards for amfepramone, cathinone, N-ethylamphetamine, fenethylline, fenproporex and mefenorex were prepared. Their purities determined by HPLC were more than 99.5%. For the identification and determination of these six drugs, their analytical data were measured and discussed by TLC, UV, IR, HPLC, GC/MS and NMR.