MilliporeSigma
  • Regio- and stereoselectivity of captodative olefins in 1,3-dipolar cycloadditions. A DFT/HSAB theory rationale for the observed regiochemistry of nitrones.
Regio- and stereoselectivity of captodative olefins in 1,3-dipolar cycloadditions. A DFT/HSAB theory rationale for the observed regiochemistry of nitrones.
The Journal of organic chemistry (2001-04-21)
R Herrera, A Nagarajan, M A Morales, F Méndez, H A Jiménez-Vázquez, L G Zepeda, J Tamariz
ABSTRACT
Captodative olefins 1-acetylvinyl carboxylates proved to be highly regioselective dipolarophiles in 1,3-dipolar cycloadditon to propionitrile oxide, arylphenylnitrile imines, diazoalkanes, and nitrones to yield the corresponding 5-substituted heterocycles. The addition of the latter was also stereoselective, being slightly susceptible to steric demand of the carboxylate substituent in the olefin. All atempts to cleave the isoxazolidine N-O bond under reductive conditions failed, providing diverse products with side-group reduction. FMO theory was unsuccessful to explain the regioselectivity observed with nitrones, since the opposite orientation was predicted. The recently formulated DFT/HSAB theoretical model was able to rationalize this regioselectivity, identifying the nucleophilic and electrophilic atoms involved in the process via calculation of interaction energies, suggesting the specific direction of the electronic process at each of the reaction sites.
Sigma-Aldrich
N-Benzylhydroxylamine hydrochloride, puriss., ≥99.0% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
In Stock
Details...
New, lower price on this item!
₩105,560
1 g
In Stock
Details...
New, lower price on this item!
₩115,507
5 g
In Stock
Details...
₩207,669
25 g
In Stock
Details...
New, lower price on this item!
₩567,588
100 g
In Stock
Details...
₩2,256,853