MilliporeSigma
  • Determination of fluquinconazole, pyrimethanil, and clofentezine residues in fruits by liquid chromatography with ultraviolet detection.
Determination of fluquinconazole, pyrimethanil, and clofentezine residues in fruits by liquid chromatography with ultraviolet detection.
Journal of AOAC International (2004-05-29)
Sandro Navickiene, Maria Lúcia Ribeiro
ABSTRACT
A simple method was developed for the determination of fluquinconazole, pyrimethanil, and clofentezine in whole fruit; peel; and pulp of mango, apple, and papaya. These compounds were extracted from fruit samples with a mixture of ethyl acetate-n-hexane (1 + 1, v/v). An aliquot (2 mL) of the extract was evaporated to near dryness under a stream of nitrogen, and the residue was dissolved with 2 mL methanol. The analysis was performed by means of liquid chromatography with ultraviolet detection at 254 nm using a gradient solvent system. The method was validated with fortified fruit samples at concentration levels of 0.05, 0.10, 0.20, and 0.50 mg/kg. Average recoveries (4-8 replicates) ranged from 80 to 95% with relative standard deviations between 3.5 and 12.7%. Detection limits ranged from 0.03 to 0.05 mg/kg for fruit pulp and 0.03 mg/kg for whole fruit. The quantitation limits ranged from 0.05 to 0.10 mg/kg for fruit pulp and 0.05 mg/kg for whole fruit. The analytical method was applied to fruit samples obtained from local markets.
Product Description
Supelco
Clofentezine, PESTANAL®, analytical standard