MilliporeSigma
  • CJZ3, a lomerizine derivative, modulates P-glycoprotein function in rat brain microvessel endothelial cells.
CJZ3, a lomerizine derivative, modulates P-glycoprotein function in rat brain microvessel endothelial cells.
Acta pharmacologica Sinica (2006-03-17)
Bian-Sheng Ji, Ling He, Xiao-Qu Li, Guo-Qing Liu
ABSTRACT
To investigate the modulatory effect of CJZ3, a lomerizine derivative, on P-glycoprotein (P-gp) function in rat brain microvessel endothelial cells (RBMEC). RBMEC were isolated and cultured in Dulbecco modified Eagle medium/F12 (1:1) medium, and the amount of intracellular rhodamine 123 (Rh123) was determined using a fluorescence spectrophotometer to evaluate the modulatory effect of CJZ3 on P-gp function. The accumulation of Rh123 was potentiated in a concentration-dependent manner after incubation with CJZ3 for RBMEC, but not for human umbilical vein endothelial cells (HUVEC). CJZ3 caused the accumulation of intracellular Rh123 in a time-dependent manner and significantly decreased the efflux of Rh123 from the cells. The inhibitory effect of CJZ3 on P-gp function was reversible and remained for 120 min after CJZ3 (2.5 micromol/L) was removed from the medium. CJZ3 has a potent in vitro effect on the inhibition of P-gp function.
Sigma-Aldrich
Lomerizine dihydrochloride, ≥98% (HPLC), powder