MilliporeSigma
  • On the production of 12 alpha-hydroxysteroid dehydrogenase from Clostridium group P, strain C48-50 ATCC 29733.
On the production of 12 alpha-hydroxysteroid dehydrogenase from Clostridium group P, strain C48-50 ATCC 29733.
Experientia (1981-05-15)
I A Macdonald
ABSTRACT
The production of 12 alpha-hydroxysteroid dehydrogenase of Clostridium group P strain C48-50 was optimized when the organism was grown in the presence of 2% fructose and 0.1% dithiothreitol. It appears that an initial redox potential of less than -160 mV (achieved by autoclaving in the presence of dithiothreitol, dithioerythritol or cysteine) is important in the production of this enzyme.
Product Description
Millipore
Brain Heart Infusion Broth, NutriSelect® Plus, according to ISO 6888, powder, for bacteria, pack of 100 g, pack of 500 g