MilliporeSigma
  • Oxidative DNA damage estimated by plasma 8-hydroxydeoxyguanosine (8-OHdG): influence of 4, 4'-methylenebis (2-chloroaniline) exposure and smoking.
Oxidative DNA damage estimated by plasma 8-hydroxydeoxyguanosine (8-OHdG): influence of 4, 4'-methylenebis (2-chloroaniline) exposure and smoking.
Journal of occupational health (2007-10-24)
Hong-I Chen, Saou-Hsing Liou, Shu-Fen Ho, Kuen-Yuh Wu, Chien-Wen Sun, Ming-Fong Chen, Li-Chuan Cheng, Tung-Sheng Shih, Ching-Hui Loh
ABSTRACT
Oxidative DNA damage may play an important role in the human carcinogenic process. Recently, we reported a case of bladder cancer among 4, 4'-methylenebis (2-chloroaniline) (MBOCA)-exposed workers. By measuring the plasma level of 8-hydroxydeoxyguanosine (8-OHdG), we investigated the association between oxidative DNA damage and MBOCA exposure. In addition, we examined the effects of different confounders on the plasma level of 8-OHdG. We undertook a cross-sectional survey at four MBOCA-producing factories in Taiwan (158 subjects). Plasma 8-OHdG levels and urinary MBOCA concentrations were measured by liquid chromatography coupled with tandem mass spectrometry (LC/MS/MS). Personal characteristics were collected by questionnaire. The workers were classified according to their job titles as exposed (n=57) or unexposed (n=101) groups as well as classified according to urinary MBOCA levels as high urinary MBOCA (>20 microg/g creatinine) (n=45) or low urinary MBOCA (n=108) groups. Neither the MBOCA-exposed workers nor the high urinary MBOCA workers had a significant increase in the mean plasma 8-OHdG level, even after adjustment for potential confounders. Age and gender were significantly positively correlated with plasma 8-OHdG levels. Smokers among high urinary MBOCA workers also had significantly higher 8-OHdG levels than non-smokers among high urinary MBOCA workers. Our study provides evidence that smoking rather than MBOCA exposure induces elevation of plasma 8-OHdG levels among workers exposed to MBOCA, indicating that oxidative DNA damage does not play an important role in the carcinogenic processes of MBOCA.
Sigma-Aldrich
4,4′-Methylene-bis(2-chloroaniline), 85%