Merck
  • In vivo kinetics of indoxyl sulfate in humans and its renal interaction with angiotensin-converting enzyme inhibitor quinapril in rats.
In vivo kinetics of indoxyl sulfate in humans and its renal interaction with angiotensin-converting enzyme inhibitor quinapril in rats.
The Journal of pharmacology and experimental therapeutics (2012-03-06)
Tomoe Fujita, Kazuhiko Ishihara, Shuichi Yasuda, Tomomi Nakamura, Mika Maeda, Mami Kobayashi, Kunihiko Sahashi, Yasuhiko Ikeda, Yuji Kumagai, Masataka Majima
ABSTRACT
Indoxyl sulfate (IS) is an organic anion uremic toxin that accumulates in patients with chronic kidney disease (CKD). The aims of this study were to examine the kinetic profiles of IS in humans at a steady state after multiple doses of L-Trp, a precursor of IS, and the in vivo interaction of IS with the angiotensin-converting enzyme inhibitor quinapril, whose active metabolite is a substrate of organic anion transporter 3 (OAT3) in rats. First, 12-h kinetics after single doses of Trp (2, 4, and 8 g) were examined in two healthy volunteers. Second, 24-h kinetics after a single dose of 2 g of Trp was studied in six volunteers. Third, 35-h kinetics after single and multiple doses of 2 g of Trp were examined in five volunteers. In anesthetized rats, quinapril or probenecid, an inhibitor of OATs, was given intravenously before IS, and blood and urine samples were taken until 90 min. Trp and IS concentrations were determined by high-performance liquid chromatography. Ultrafiltration was used to measure serum unbound IS concentrations. Renal tubular secretion of IS accounted for more than 90% of its renal clearance in the steady state of serum IS levels after multiple doses in humans. In animals, the serum area under the curve of IS increased in conjunction with a decrease in renal clearances after coadministration of IS with quinapril or probenecid. It is concluded that quinapril may inhibit the urine excretion of IS via OAT3-mediated renal tubular transport in patients with CKD.
Sigma-Aldrich
Quinapril hydrochloride, ≥98% (HPLC), solid
Pricing and availability is not currently available.