MilliporeSigma
  • Identification of food-derived elastin peptide, prolyl-glycine (Pro-Gly), in human blood after ingestion of elastin hydrolysate.
Identification of food-derived elastin peptide, prolyl-glycine (Pro-Gly), in human blood after ingestion of elastin hydrolysate.
Journal of agricultural and food chemistry (2012-05-03)
Yasutaka Shigemura, Misako Nakaba, Eri Shiratsuchi, Masashi Suyama, Michio Yamada, Tamami Kiyono, Kazuna Fukamizu, Eun Young Park, Yasusi Nakamura, Kenji Sato
ABSTRACT
Elastin hydrolysate has apparent beneficial effects, and the food-derived peptide prolyl-glycine (Pro-Gly) is present in human blood after oral ingestion. Following ingestion of elastin hydrolysate (10 g/60 kg body weight) by healthy human volunteers, peripheral blood was used to prepare plasma samples from which peptides were extracted by solid phase extraction and fractionated by size-exclusion chromatography (SEC). Peptides in the SEC fractions were derivatized with phenyl isothiocyanate (PITC) and resolved by reversed phase (RP)-HPLC. Pro-Gly was the major food-derived elastin peptide, reaching a maximum (18 μM) at 30 min after ingestion, and decreasing to approximately 20% at 4 h after ingestion. Finally, in cell culture, levels of Pro-Gly in the medium above 0.1 μg/mL significantly enhanced elastin synthesis of normal human dermal fibroblasts (NHDF) without affecting the rate of cell proliferation.
Sigma-Aldrich
Pro-Gly