MilliporeSigma
  • Ganglioside GD2 identifies breast cancer stem cells and promotes tumorigenesis.
Ganglioside GD2 identifies breast cancer stem cells and promotes tumorigenesis.
The Journal of clinical investigation (2012-05-16)
Venkata Lokesh Battula, Yuexi Shi, Kurt W Evans, Rui-Yu Wang, Erika L Spaeth, Rodrigo O Jacamo, Rudy Guerra, Aysegul A Sahin, Frank C Marini, Gabriel Hortobagyi, Sendurai A Mani, Michael Andreeff
ABSTRACT
Cancer stem cells (CSCs) are a small subpopulation of cancer cells that have increased resistance to conventional therapies and are capable of establishing metastasis. However, only a few biomarkers of CSCs have been identified. Here, we report that ganglioside GD2 (a glycosphingolipid) identifies a small fraction of cells in human breast cancer cell lines and patient samples that are capable of forming mammospheres and initiating tumors with as few as 10 GD2+ cells. In addition, the majority of GD2+ cells are also CD44hiCD24lo, the previously established CSC-associated cell surface phenotype. Gene expression analysis revealed that GD3 synthase (GD3S) is highly expressed in GD2+ as well as in CD44hiCD24lo cells and that interference with GD3S expression, either by shRNA or using a pharmacological inhibitor, reduced the CSC population and CSC-associated properties. GD3S knockdown completely abrogated tumor formation in vivo. Also, induction of epithelial-mesenchymal transition (EMT) in transformed human mammary epithelial cells (HMLER cells) dramatically increased GD2 as well as GD3S expression in these cells, suggesting a role of EMT in the origin of GD2+ breast CSCs. In summary, we identified GD2 as a new CSC-specific cell surface marker and GD3S as a potential therapeutic target for CSCs, with the possibility of improving survival and cure rates in patients with breast cancer.
Sigma-Aldrich
Disialoganglioside-GD2 from bovine brain, ~95%, lyophilized powder, semisynthetic