MilliporeSigma
  • Systematic study of the physicochemical properties of a homologous series of aminobisphosphonates.
Systematic study of the physicochemical properties of a homologous series of aminobisphosphonates.
Molecules (Basel, Switzerland) (2012-09-14)
Aino-Liisa Saari, Helena Hyvönen, Manu Lahtinen, Markku Ylisirniö, Petri Turhanen, Erkki Kolehmainen, Sirpa Peräniemi, Jouko Vepsäläinen
ABSTRACT
Aminobisphosphonates, e.g., alendronate and neridronate, are a well known class of molecules used as drugs for various bone diseases. Although these molecules have been available for decades, a detailed understanding of their most important physicochemical properties under comparable conditions is lacking. In this study, ten aminobisphosphonates, H(2)N(CH2)(n)C(OH)[P(O)(OH)(2)](2) in which n = 2-5, 7-11 and 15 have been synthesized. Their aqueous solubility as a function of temperature and pH, pK(a-) values, thermal stability, IR absorptions, and NMR spectral data for both liquid (1)H, (13)C, (31)P-NMR and solid state(13)C, (15)N and (31)P-CPMAS NMR) were determined.
Sigma-Aldrich
Neridronate, ≥97% (NMR), solid