MilliporeSigma
  • Rapid imaging, using a cooled charge-coupled-device, of fluorescent two-dimensional polyacrylamide gels produced by labelling proteins in the first-dimensional isoelectric focusing gel with the fluorophore 2-methoxy-2,4-diphenyl-3(2H)furanone.
Rapid imaging, using a cooled charge-coupled-device, of fluorescent two-dimensional polyacrylamide gels produced by labelling proteins in the first-dimensional isoelectric focusing gel with the fluorophore 2-methoxy-2,4-diphenyl-3(2H)furanone.
Electrophoresis (1988-07-01)
P Jackson, V E Urwin, C D Mackay
ABSTRACT
A new method for visualising proteins in two-dimensional polyacrylamide gels was developed. Proteins were labelled with the fluorophore 2-methoxy-2,4-diphenyl-3(2H)furanone (MDPF) while present in the first-dimensional gel after isoelectric focusing and subsequently electrophoresed into the second-dimensional gel. High resolution spot patterns were produced and compared with other methods of visualisation. A new rapid imaging system based on a cooled charge-coupled-device was used to view the two-dimensional fluorescent protein spot patterns. The method allows the immediate and rapid imaging of two-dimensional gels at the end of electrophoresis with no further processing.
Sigma-Aldrich
2-Methoxy-2,4-diphenyl-3(2H)-furanone, suitable for fluorescence, ≥98.0% (HPLC)