MilliporeSigma
  • [Pharmacologic evaluation of the feasibility of cartilage therapy in degenerative joint diseases (arthroses)].
[Pharmacologic evaluation of the feasibility of cartilage therapy in degenerative joint diseases (arthroses)].
Zeitschrift fur Rheumatologie (1983-07-01)
D A Kalbhen
ABSTRACT
Due to their pharmacological properties most antiphlogistic/antirheumatic drugs are successfully used for treatment of inflammatory rheumatic diseases, but they are not able to counteract cartilage degeneration in osteoarthrosis. For specific therapy of osteoarthrosis only those drugs are suitable, which are able to inhibit enzymatic breakdown of articular cartilage, stimulate anabolic processes in cartilage, and to enhance the supply of nutritional and energy substrates for the cartilage cells. In this respect drugs such as Arteparon, Rumalon, Dona 200-S, Glyvenol or Pentosan polysulfate are of interest. A great number of pharmacological experiments have shown, that certain chondroprotective agents exert pronounced anti-degenerative effects, which can be quantitatively demonstrated in laboratory animals within 3 to 4 months by macroscopical, radiological and histological methods. Additional biochemical and in-vitro studies have elucidated some interesting chondro-protective, chondro-stimulatory or chondro-nutritive properties of certain drugs. In agreement with our experimental results clinical experiences have proved the efficacy of chondro-protective agents. Due to the bradytrophia of human articular cartilage a chondro-protective therapy may be only effective in long-term applications, resulting in a significant reduction of the intensity and progression of the degenerative joint destruction.
Product Description
Tribenoside, European Pharmacopoeia (EP) Reference Standard