MilliporeSigma
  • Fatty acyl derivatives of glucosamine 1-phosphate in Escherichia coli and their relation to lipid A. Complete structure of A diacyl GlcN-1-P found in a phosphatidylglycerol-deficient mutant.
Fatty acyl derivatives of glucosamine 1-phosphate in Escherichia coli and their relation to lipid A. Complete structure of A diacyl GlcN-1-P found in a phosphatidylglycerol-deficient mutant.
The Journal of biological chemistry (1983-06-25)
K Takayama, N Qureshi, P Mascagni, M A Nashed, L Anderson, C R Raetz
ABSTRACT
We have determined the complete structure of a glycolipid (designated lipid X) previously found to accumulate in certain Escherichia coli mutants defective in phosphatidylglycerol synthesis (Nishijima, M., and Raetz, C.R.H. (1979) J. Biol. Chem. 254, 7837-7844). Based on fast atom bombardment mass spectrometry and proton nuclear magnetic resonance studies, this substance is an acylated metabolite of glucosamine 1-phosphate. Lipid X of E. coli has a Mr = 711.87 as the free acid (C34H66NO12P) and contains two beta-hydroxymyristate moieties, one attached as an amide at the 2 position and the other as an ester at the 3 position of the sugar. It has free hydroxyl groups at the 4 and 6 positions, and the anomeric configuration is alpha. The structure of lipid X from E. coli closely resembles the reducing end subunit of lipid A, and it might represent a very early precursor in the biosynthesis of lipid A. To our knowledge, fatty acyl derivatives of glucosamine 1-phosphate have not been reported previously.
Sigma-Aldrich
α-D-Glucosamine 1-phosphate