MilliporeSigma
  • Bioassay for carcinogenicity of 3,2'-dimethyl-4-nitrosobiphenyl, O-nitrosotoluene, nitrosobenzene and the corresponding amines in Syrian golden hamsters.
Bioassay for carcinogenicity of 3,2'-dimethyl-4-nitrosobiphenyl, O-nitrosotoluene, nitrosobenzene and the corresponding amines in Syrian golden hamsters.
Cancer letters (1983-10-01)
S S Hecht, K El-Bayoumy, A Rivenson, E S Fiala
ABSTRACT
3,2'-Dimethyl-4-aminobiphenyl and 3,2'-dimethyl-4-nitrosobiphenyl were administered by subcutaneous injection in peanut oil to 2 groups of 15 male and 15 female Syrian golden hamsters. The total dose of each compound was 5.6 mmol/kg. In the group treated with 3,2'-dimethyl-4-aminobiphenyl, 24 animals had bladder tumors. In the group treated with 3,2'-dimethyl-4-nitrosobiphenyl 25 animals had subcutaneous tumors and 2 had bladder tumors. These results indicate that 3,2'-dimethyl-4-nitrosobiphenyl is a potent locally acting carcinogen. Total doses of 99 mmol/kg of aniline, o-toluidine, nitrosobenzene, or o-nitrosotoluene administered by subcutaneous injection failed to induce tumors.
Sigma-Aldrich
2-Nitrosotoluene, 97%