MilliporeSigma
  • Formation of the thioester, N,S-diacetylcysteine, from acetaldehyde and N,N'diacetylcystine in aqueous solution with ultraviolet light.
Formation of the thioester, N,S-diacetylcysteine, from acetaldehyde and N,N'diacetylcystine in aqueous solution with ultraviolet light.
Journal of molecular evolution (1981-01-01)
A L Weber
ABSTRACT
The thioester, N,S-diacetylcysteine, is formed during the illumination of phosphate buffered (pH 7.0) aqueous solutions of acetaldehyde and N,N'-diacetylcystine with ultraviolet light. The yield of N,S-diacetylcysteine relative to N-acetylcysteine and unidentified products progressively increases as ultraviolet light below 239 nm, 253 nm and 281 nm is cut off with optical filters. When ultraviolet light below 320 nm is removed with an optical filter, there is no detectable reaction. Illumination of 0.025 M N,N'-diacetylcystine with 0.5 M and 1.0 M acetaldehyde with filtered ultraviolet light gives, respectively, 20% and 80% yields of N,S-diacetylcysteine. In the reaction with 1.0 M acetaldehyde, N-acetylcysteine forms early in the reaction and later decreases with its conversion to N,S-diacetylcysteine. The prebiotic significance of these reactions is discussed.
Product Description
Acetylcysteine impurity D, European Pharmacopoeia (EP) Reference Standard