MilliporeSigma
  • Activation of the macrophage respiratory burst by phorbol myristate acetate: evidence for both tyrosine-kinase-dependent and -independent pathways.
Activation of the macrophage respiratory burst by phorbol myristate acetate: evidence for both tyrosine-kinase-dependent and -independent pathways.
Biochimica et biophysica acta (1994-06-30)
S P Green, W A Phillips
ABSTRACT
We have investigated the relationship between tyrosine phosphorylation and respiratory-burst activity in murine bone-marrow-derived macrophages (BMM) stimulated with phorbol myristate acetate (PMA). In unprimed BMM, a good correlation was observed between the net level of tyrosine phosphorylation and the activity of the respiratory burst. The phosphotyrosine phosphatase inhibitor, vanadate, enhanced both tyrosine phosphorylation and respiratory-burst activity triggered by PMA. Furthermore, the tyrosine kinase inhibitor, ST638, abolished both tyrosine phosphorylation and respiratory-burst activity stimulated by PMA. However, in BMM primed by preexposure to TNF alpha, the correlation between net tyrosine phosphorylation and respiratory-burst activity triggered by PMA was not maintained. ST638 was found to only partially inhibit the PMA-triggered respiratory burst under conditions where PMA-stimulated tyrosine phosphorylation was abolished. We conclude that PMA can activate the macrophage respiratory burst by both tyrosine-kinase-dependent and -independent pathways.
Sigma-Aldrich
ST638, ≥98% (HPLC), solid