MilliporeSigma
  • Evidence for 5-HT1-like receptor-mediated vasoconstriction in human pulmonary artery.
Evidence for 5-HT1-like receptor-mediated vasoconstriction in human pulmonary artery.
British journal of pharmacology (1996-09-01)
M R MacLean, R A Clayton, A G Templeton, I Morecroft
ABSTRACT
1. The 5-hydroxytryptamine (5-HT) receptors mediating contraction of human isolated pulmonary artery rings were investigated. Responses to the agonists 5-carboximidotryptamine (5-CT, non-selective 5-HT1 agonist), sumatriptan (5-HT1D-like receptor agonist), 5-HT and 8-hydroxy-2-(di-n-propylamino)-tetralin (8-OH-DPAT, 5-HT1A receptor agonist) were studied. Responses to 5-HT and sumatriptan in the presence of the antagonists, methiothepin (non-selective 5-HT1+2-receptor antagonist), ketanserin (5-HT2A receptor antagonist) and the novel antagonist, GR55562 (5-HT1D receptor antagonist) were also studied. 2. All agonists contracted human pulmonary artery ring preparations in the following order of potency 5-CT > 5-HT = sumatriptan > 8-OH-DPAT. Maximum responses to 5-HT, 5-CT and sumatriptan were not significantly different. 3. Methiothepin 1 nM and 10 nM, but not 0.1 nM reduced the maximum contractile responses to 5-HT but did not alter tissue sensitivity to 5-HT. Methiothepin 0.1 nM, 1 nM and 10 nM had a similar effect on responses to sumatriptan. 4. The 5-HT2A receptor antagonist ketanserin (10 nM, 100 nM and 1 microM) also reduced the maximum contractile response to both 5-HT and sumatriptan without affecting tissue sensitivity to these agonists. 5. The novel 5-HT1D receptor antagonist, GR55562, inhibited responses to 5-HT and sumatriptan in a true competitive fashion. 6. The results suggest that the human pulmonary artery has a functional population of 5-HT1D-like receptors which are involved in the contractile response to 5-HT.