MilliporeSigma
  • Prevention of co-elution of steroid sulfates with serum proteins from pre-column in column-switching HPLC system.
Prevention of co-elution of steroid sulfates with serum proteins from pre-column in column-switching HPLC system.
Biological & pharmaceutical bulletin (1998-12-16)
N Tagawa, H Tsuruta, A Fujinami, Y Kobayashi
ABSTRACT
A method to prevent co-elution of steroid sulfates with proteins in serum from the pre-column in column-switching HPLC was developed. The pre-column, a polymer-coated mixed function column, was used for ion-pair chromatography with 5 mM tetra-n-butylammonium (TBA) ion. As steroid sulfates, estriol 3-sulfate, dehydroepiandrosterone 3-sulfate and pregnenolone 3-sulfate were used. Human serum (25 microl) was diluted with mobile phases including 5, 100 and 500 mM TBA ion, and then injected directly into the pre-column. The peak areas of the steroid sulfates in serum samples were compared with those of the steroid standards without serum. When 25/microl of serum was diluted with mobile phase including 100 or 500 mM TBA ion, the steroid sulfates in serum were retained in the pre-column; however, the steroid sulfates from the same sample diluted with mobile phase containing 5 mM TBA ion were not retained in the pre-column. Addition of an excess amount of counter ion (TBA ion) into the serum sample made it possible to retain the steroid sulfates in the pre-column. This method was applied to column-switching HPLC for measurement of steroid sulfates in serum using a semi-microcolumn as the analytical column.
Sigma-Aldrich
Estriol 3-sulfate sodium salt, ≥98% (TLC)