MilliporeSigma
  • An inherited metabolic disorder presenting as ethylene glycol intoxication in a young adult.
An inherited metabolic disorder presenting as ethylene glycol intoxication in a young adult.
The American journal of forensic medicine and pathology (2002-04-16)
Karen Pien, Bruno van Vlem, Rudy van Coster, Georges Dacremont, Michel Piette
ABSTRACT
Despite the abundance of reports emerging in the literature on metabolic disorders, some disorders remain undiagnosed or misdiagnosed, not only in clinical pathology but also in forensic pathology. The authors report a patient who had recurrent episodes characterized by nausea, vomiting, and signs of dehydration necessitating admission to the hospital. At each admission, he was found to have lactic acidosis. On the first admission, glycolic acid was detected in his blood and he was diagnosed as having ethylene glycol intoxication. Only at the third admission, 2 years after the first, was the possibility of an underlying metabolic disorder considered. Laboratory investigations showed a deficiency of complex I in the mitochondrial oxidative phosphorylation. Possible medicolegal implications are discussed.
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00