MilliporeSigma
Blood transfusions in the therapy of a case of prolidase deficiency.
The British journal of dermatology (1992-02-01)
E Berardesca, D Fideli, M Bellosta, K M Dyne, G Zanaboni, G Cetta
ABSTRACT
A case is reported of a 15-year-old boy with prolidase deficiency and marked urinary excretion of the iminodipeptide gly-pro. Prolidase activity of erythrocytes against substrate glycyl-proline was deficient, but after blood transfusions this was increased to 15.7% of donor activity and declined to 12% and 3.4% of normal activity after 8 and 45 days, respectively. Urinary iminodipeptide levels following transfusion remained unaltered.
Sigma-Aldrich
Gly-Pro